Dulbecco's Modification of Eagle's Medium, MP Biomedicals
Fournisseur: MP Biomedicals
091233454
ICNA091233454EA 48.6 EUR InStock
ICNA091233454
Dulbecco's Modification of Eagle's Medium, MP Biomedicals
Milieux de culture cellulaire
Dulbecco's Modification of Eagle's Medium with 20 mM HEPES and 4,5 g Dextrose/liter, without L-glutamine or Sodium bicarbonate is ideally suited for supporting and maintaining a vast range of mammalian cell types.
  • Most widely used modification of Eagle's Basal Medium (BME)
  • Contains four times greater concentration of amino acids, vitamins and supplementary components
  • For optimal culturing of other cell types, a modification of 4500 mg/L glucose is available
The original formulation calls for 1000 mg/L of glucose, which was first employed to support the polyoma virus in primary and secondary embryonic mouse cultures.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité