Dulbecco's Modification of Eagle's Medium (1X), MP Biomedicals
Fournisseur: MP Biomedicals
091642954
ICNA091642954EA 64.8 EUR InStock
ICNA091642954
Dulbecco's Modification of Eagle's Medium (1X), MP Biomedicals
Milieux de culture cellulaire
Dulbecco's Modification of Eagle's Medium 1X with 4.5 g/L glucose, without L-glutamine, i-inositol is ideally suited for supporting and maintaining a vast range of mammalian cell types.
  • Most widely used modification of Eagle's Basal Medium (BME)
  • Contains four times greater concentration of amino acids, vitamins and supplementary components
The original formulation calls for 1000 mg/L of glucose, which was first employed to support the polyoma virus in primary and secondary embryonic mouse cultures.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité