Tryptose Phosphate Broth, MP Biomedicals
Fournisseur: MP Biomedicals
091682149
ICNA091682149EA 47.3 EUR InStock
ICNA091682149
Tryptose Phosphate Broth, MP Biomedicals
Milieux de culture cellulaire
Tryptose Phospate Broth (TPB) has been studied as supplement for the preparation of media that supports vaccine production in BHK-21 cells and the growth of SF21 insect cells in high-density perfusion culture stirred-tank bioreactors.
  • Used for testing the sensitivity of microorganims to antibiotics by the tube dilution method
  • Used for cultivating fastidious microorganism
To rehydrate the powdered medium, dissolve 29.5 grams in one liter of water. Dispense as desired. Sterilize in the autoclave for 15 minutes at 15 pounds pressure at 121 °C (Catalog number 16821 is a pre-sterilized solution). For more rapid growth of pathogenic bacteria, dissolve one gram of agar per liter of broth prior to sterilizing the medium. Final pH will be approximately 7.3 ±0.2 at 25 °C. The addition of 0.1 to 0.2% agar to a broth medium improves the productivity of the medium for most purposes. The low agar concentration provides suitable conditions for both aerobic growth in the clear upper zone and for microaerophilic and anaerobic growth in the lower, flocculent agar zones.
Diagnostic Procedures and Reagents (APHA) also recommended adding 0.1 to 0.2% agar to the formulation. While studying the detection of food poisoning attributable to dairy products, Newman used tryptose phosphate broth with 0.1% agar and 1:2500 sodium azide added for cultivating streptococci at 37 °C.
Waisbren, Carr and Dunnett demonstrated that a soy bean peptone medium inhibited the action of neomycin, Aureomycin®, terramycin and polymyxin against the test organism while tryptose phosphate broth, dextrose broth and nutrient broth were suitable for the test.
Standard Methods for the Examination of Dairy Products, 9th Edition, described tryptose phosphate agar broth (0.1% agar added) for use in isolating pathogenic bacteria from cheese.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité