Newborn Bovine Serum, MP Biomedicals
Fournisseur: MP Biomedicals
092912154
ICNA092912154EA 150 EUR InStock
ICNA092912154
Newborn Bovine Serum, MP Biomedicals
Sérums
This serum is produced from blood collected from newly born calves between 1 to 14 days of age.
  • Cell culture and organ preservation
  • Culture Media
  • Suitable for cell culture
  • Shelf Life 36 months
Newborn Bovine Serum (NBS) is useful in a wide range of cell culture and organ preservation applications. For example, it is used in cornea preservation and cryopreservation of bovine embryos. NBS is used in cell culture to support the maturation of oocytes, to study the function of tubular epithelial cells, and to develop primary cultures of cells such asMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité