Dehydrated culture media, Terrific broth II, MP Biomedicals
Fournisseur: MP Biomedicals
113046075
ICNA113046075EA 235 EUR InStock
ICNA113046075
Dehydrated culture media, Terrific broth II, MP Biomedicals
Milieux pour microbiologie Milieux prêts à l'emploi de microbiologie
Terrific broth (TB) has been formulated for optimum growth of Escherichia coli for purposes of strain maintenance, cloning, protein expression. It is certified for plasmid and other bacterial cultures in liquid medium. Higher cell densities are achieved using this broth.
  • Molecular biology certified bacterial medium
Terrific Broth II is a proprietary formulation that is shipped as a single powder instead of as two separate components exhibiting same growth characteristics as the original terrific broth formula.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité