Genomic RNA purification kits, Fast RNA™ Pro Blue kit, MP Biomedicals
Fournisseur: MP Biomedicals
116025050
ICNA116025050EA 286 EUR InStock
ICNA116025050
Genomic RNA purification kits, Fast RNA™ Pro Blue kit, MP Biomedicals
Réactifs pour acides nucléiques Nucleic Acid Purification Kits and Reagents
FastRNA® Pro Blue Kit is used to isolate total RNA from gram positive and gram negative bacteria. Isolated RNA is suitable for RT-PCR analysis and other downstream applications.
Using the FastPrep®-24 or FastPrep® FP120 instrument, up to 10¹⁰ cells are homogenised by Lysing Matrix B in impact-resistant 2 ml tubes. Total RNA is released into the proprietary, protective RNApro™ Solution.
For research purposes only.
Information de livraison: Kit contains RNApro™ Solution (1×55 ml), DEPC-treated H₂O (1×15 ml) and Lysing Matrix B (50×2 ml tubes).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité