Fast RNA™ spin kit for yeast, MP Biomedicals
Fournisseur: MP Biomedicals
116030050
ICNA116030050EA 456 EUR InStock
ICNA116030050
Fast RNA™ spin kit for yeast, MP Biomedicals
Réactifs pour acides nucléiques Nucleic Acid Purification Kits and Reagents
The FastRNA™ SPIN Kit for Yeast quickly and efficiently isolates high-quality, total RNA from tough-to-lyse yeast strains, fungi, and algae in approximately 15 minutes using a specialised Lysing Matrix (Yttria-stabilised zirconium oxide, 0,5 mm) for cell lysis and SPIN columns for the purification process.
The FastRNA™ SPIN Kit isolates both large and small RNA species without the use of phenol or reducing agents. Small RNAs (eg. tRNAs, microRNAs) can be recovered following a simple adjustment within the RNA isolation protocol. RNA (~5 µg) isolated from 10¹ to 10⁵ cells can be eluted into volumes as little as 6 µl in less than 15 minutes. High quality total RNA (A260/A280 >1,8, A260/A230 >1,8) is recovered. RNA is eluted with RNase-free water and can be stored at –70 °C. The addition of RNase inhibitors is optional, but highly recommended for prolonged storage.
For research purposes only.
Information de livraison: Kit contains Lysing Matrix Y (0,5 mm Yttria-stabilised ZrO2), 50×2 ml tubes; FastRNA™ Lysis Buffer, 50 ml; FastRNA™ Prep Buffer, 25 ml; FastRNA™ Wash Buffer, 12 ml; Water (DNase/RNase-free), 1 ml; FastRNA™ Spin Column 1, 50 each; FastRNA™ Spin Column 2, 50 each; and Collection Tubes (2 ml), 100 each.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité