Genomic RNA purification kits, Fast RNA™ Pro Red kit, MP Biomedicals
Fournisseur: MP Biomedicals
116035050
ICNA116035050EA 272 EUR InStock
ICNA116035050
Genomic RNA purification kits, Fast RNA™ Pro Red kit, MP Biomedicals
Réactifs pour acides nucléiques Nucleic Acid Purification Kits and Reagents
FastRNA® Pro Red Kit is used to isolate total RNA from yeast and fungi. Isolated RNA is suitable for RT-PCR analysis and other downstream applications
Using the FastPrep-24™ or FastPrep® FP120 instrument, up to 10⁹ cells are homogenised by Lysing Matrix C in impact-resistant 2 ml tubes. Total RNA is released into the proprietary, protective RNApro™ Solution. Following extraction with chloroform and precipitation, total RNA is extracted.
For research purposes only.
Information de livraison: Kit contains RNApro™ Solution (1×55 ml), DEPC-treated H₂O (1×15 ml) and Lysing Matrix C (50×2 ml tubes).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité