Fast RNA™ Pro green kit, MP Biomedicals
Fournisseur: MP Biomedicals
116045050
ICNA116045050EA 286 EUR InStock
ICNA116045050
Fast RNA™ Pro green kit, MP Biomedicals
Réactifs pour acides nucléiques Nucleic Acid Purification Kits and Reagents
FastRNA® Pro Green Kit is used to isolate total RNA from any type of plant and animal tissue or cultured cells. Extracted RNA is suitable for RT-PCR analysis and other downstream applications.
The FastRNA™ Pro Green Kit is designed to efficiently isolate total RNA from any type of plant and animal tissue or cultured cells. Using the FastPrep-24™ or FastPrep® FP120 instrument, between 50 to 500 mg of tissue can be homogenised by Lysing Matrix D in impact-resistant 2 ml tubes. Total RNA is released into the proprietary, protective RNApro™ Solution. Following extraction with chloroform and precipitation, total RNA is extracted.
For research purposes only.
Information de livraison: Kit contains RNApro™ Solution, 1×55 ml; DEPC-H2O, 1×15 ml; and Lysing Matrix D, 50×2 ml tubes.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité