Fast RNA™ Pro Soil-Indirect Kit, MP Biomedicals
Fournisseur: MP Biomedicals
116075050
ICNA116075050EA 589 EUR InStock
ICNA116075050
Fast RNA™ Pro Soil-Indirect Kit, MP Biomedicals
Réactifs pour acides nucléiques Nucleic Acid Purification Kits and Reagents
FastRNA™ Pro Soil-Indirect Kit is used to isolate total RNA from organic material found in soil samples. This kit allows for the sample to be slurried with water or buffer in order to further separate biomass from the soil or environmental matrix, and the resulting supernatant is then filtered, lysed, and purified through the rest of the protocol. Total RNA isolated is suitable for RT-PCR analysis and other downstream applications.
The FastRNA™ Pro Soil-Indirect Kit is designed to efficiently isolate total RNA from organic material found in soil supernatants. Cellular material is washed away from soil particles, centrifuged, and homogenised by any FastPrep® instrument in impact-resistant 2 ml tubes of Lysing Matrix E. Total RNA is released into a protective solution, extracted, precipitated, and cleaned with the proprietary RNAMATRIX™ and a Quick-Clean Spin Filter. Total RNA prepared with this kit is suitable for RT-PCR analysis and other downstream applications.
For research purposes only.
Information de livraison: Kit contains RNApro™ Solution; Lysing Matrix E; Inhibitor Removal Solution; DEPC-treated H₂O; RNAMATRIX® Binding Solution; RNAMATRIX® Slurry; RNAMATRIX® Wash Solution; and Quick-Clean Spin Filters.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité