Genomic DNA purification kits, Fast DNA™ SPIN kit for Soil, MP Biomedicals
Fournisseur: MP Biomedicals
116560600
ICNA116560600EA 243 EUR InStock
ICNA116560600
Genomic DNA purification kits, Fast DNA™ SPIN kit for Soil, MP Biomedicals
Réactifs pour acides nucléiques Nucleic Acid Purification Kits and Reagents
The FastDNA™ SPIN Kit for Soil is designed to efficiently isolate bacterial, fungi, plant and animal genomic DNA from soil and environmental samples.
Up to 5 g of soil are processed by the FastPrep®-24 or FastPrep®-96 instrument with the Lysing Matrix E tubes designed to efficiently lyse all microorganisms, including difficult sources such as eubacterial spores and endospores, gram positive bacteria and yeast. The released DNA is purified by a silica-based spin filter columns and removes humic acids/polyphenols that can inhibit PCR. The resulting pure DNA is suitable for PCR analysis and other downstream applications.
Information de livraison: Garnet lysing matrix tubes (50 ml), 10 each; PLS solution, 60 ml; MT buffer, 15 ml; sodium phosphate buffer, 350 ml; PPS for soil, 25 ml; binding matrix, 120 ml; GTC wash buffer, 120 ml; SEWS-M, 12 ml; TES, 60 ml; BBS gel loading dye, 200 µl; SPIN filter tubes (50 ml), 10 each.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité