Bovine serum albumin (BSA), fraction V, MP Biomedicals
Fournisseur: MP Biomedicals
16006910
ICNA16006910EA 52.85 EUR InStock
ICNA16006910
Bovine serum albumin (BSA), fraction V, MP Biomedicals
Protéines et Peptides
Albumins are a group of acidic proteins which occur in the body fluids and tissues of mammals and in some plant seeds.
  • Readily soluble in water
It is a general blocking reagent in ELISA, as a nutrient in cell and microbial culture, it also stabilise some enzymes during digestion of DNA. It is used to determine the quantity of other proteins, by comparing an unknown quantity of protein to known amounts of BSA.
Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité