Immersion Circulators, ICC 150 lite pro 9
Fournisseur: IKA
10013199
IKAA10013199EA 1900 EUR InStock
IKAA10013199
Immersion Circulators, ICC 150 lite pro 9
Thermostats à circulation Systèmes à circulation ouverte Contrôleurs à thermostat
The compact immersion circulators of the lite series are powerful and of high quality, with a focus on essential basic functions, excellent performance and efficient operation.
  • Max. temperature in stainless steel bath (pro): 150 °C
ICC 150 lite is suitable for tempering liquids up to 150 °C. The integrated pressure/suction pump allows ICC 150 lite to be used for internal and external applications.
To make configuration of your temperature control system as convenient as possible, IKA offers 'ready-to-connect' packages with all the necessary accessories.
Information de livraison: Package includes: ICC 150 lite compact immersion circulator, IB 9 pro stainless steel bath (size S), BS.ICC lite bath bridge, small. Technical data relates to the temperature control instrument included in the package.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité