Immersion Circulators, ICC 150 lite pro 20
Fournisseur: IKA
10013221
IKAA10013221EA 2120 EUR InStock
IKAA10013221
Immersion Circulators, ICC 150 lite pro 20
Thermostats à circulation Systèmes à circulation ouverte Contrôleurs à thermostat
The compact immersion circulators of the lite series are powerful and of high quality, with a focus on essential basic functions, excellent performance and efficient operation.
  • Max. temperature in stainless steel bath (pro): 150 °C
ICC 150 lite is suitable for tempering liquids up to 150 °C. The integrated pressure/ suction pump allows ICC 150 lite to be used for internal and external applications.
To make configuration of your temperature control system as convenient as possible, IKA offers 'ready-to-connect' packages with all the necessary accessories.
Information de livraison: Package includes: ICC 150 lite compact immersion circulator, IB 20 pro stainless steel bath (size L), BL.ICC lite bath bridge, big. Technical data relates to the temperature control instrument included in the package.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité