UTTD Control Life Science Solution Homogenising System
Fournisseur: IKA
10013296
IKAA10013296EA 1708 EUR InStock
IKAA10013296
UTTD Control Life Science Solution Homogenising System
Homogénéisateurs Homogénéisateurs de paillasse
The UTTD control life science solution package was specially put together to meet the critical challenges of homogenising tissue, cartilage and bone samples: homogeneous comminution of the samples and protection against cross-contamination.
  • Single-use homogenisation of tissue samples without risk of cross-contamination
The package includes the powerful ULTRA-TURRAX® Tube Drive P control homogenisation system, which has been designed for universal use. In combination with the patented DT-20 disposable dispersing chambers with 20 ml volume and rotor-stator element, this system ensures optimum sample comminution and fast and uniform homogenisation of the samples.
The hermetically sealable disposable sample containers serve simultaneously as a dispersion vessel, storage unit and transport container. They significantly reduce the cleaning effort and systematically eliminate cross-contamination. Users benefit from reduced handling costs and a lower number of incorrect analyses. Samples can be collected or media added via the pierceable membrane in the vessel lid.
Information de livraison: ULTRA-TURRAX® Tube Drive P control, DT-20 Dispersing tube, 20 ml (25 pieces.)
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité