Tube Mill Control Agriculture Solution Package
Fournisseur: IKA
10013298
IKAA10013298EA 2419 EUR InStock
IKAA10013298
Tube Mill Control Agriculture Solution Package
Broyeurs
Tube mill control agriculture solution system is used for grinding of seeds and grains without risk of cross-contamination.
The Tube Mill control agriculture solution package, consist of the tube mill control batch mill and patented disposable plastic chambers with a volume of 40 ml.
Grinding, storage and transport of the samples takes place in a closed, disposable grinding chamber which eliminates the risk of cross-contamination between different batches. The time-consuming cleaning of the mill and the containers after each cycle is also eliminated. The MT grinding chambers are designed to process even the smallest sample quantities from 1 g.
The Tube Mill control is a batch mill for grinding hard, brittle and dry materials up to Mohs hardness 5. With its disposable and reusable grinding chambers with a volume of 40 ml, it is versatile and can be used for a wide range of requirements.
Information de livraison: Tube mill control, MT 40.10 Disposable grinding chamber (10 pieces.), MMT 40.1 Multi-use milling tube.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité