ROT.ROTATEMP lo-vi Viscometer Package
Fournisseur: IKA
10014004
IKAA10014004EA 6936 EUR InStock
IKAA10014004
ROT.ROTATEMP lo-vi Viscometer Package
Viscosimètres Viscosimètres rotatifs
The ROTATEMP lo-vi package is the ideal solution for precise and reliable viscosity measurement of difficult-to-measure, highly viscous samples.
  • Fast, precise and reproducible results: The combination of ROTAVISC viscometer and ROTATEMP high-temperature system simplifies the viscosity measurement of difficult-to-measure, highly viscous samples
  • Simple operation and space-saving: The complete system is supplied ready for connection. With its space-saving design, it fits easily into any laboratory
  • Optimum compatibility and automation: ROTAVISC and ROTATEMP
More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité