Immersion Circulators, ICC 100 lite
Fournisseur: IKA
20105647
IKAA20105647EA 902 EUR InStock
IKAA20105647
Immersion Circulators, ICC 100 lite
Thermostats à circulation Systèmes à circulation ouverte Contrôleurs à thermostat
The compact immersion circulators of the lite series are powerful and of high quality, with a focus on essential basic functions, excellent performance and efficient operation. ICC 100 lite is suitable for tempering liquids up to 100 °C and internal circulation.
  • USB and RS232 interface for monitoring, control and documentation
Increased safety: ICC 100 lite is approved for use with flammable liquids in accordance with DIN 12876, safety class III.
Convenient handling: The integrated stand enables easy cleaning and prevents important components from being damaged.
Durability and low maintenance: Robust design thanks to high-quality material, housing made of coated stainless steel.
Easy configuration: To make configuration of your temperature control system as convenient as possible, IKA offers 'ready-to-connect' packages with all the necessary accessories.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité