ROTATEMP High-Temperate System
Fournisseur: IKA
20113825
IKAA20113825EA 3800 EUR InStock
IKAA20113825
ROTATEMP High-Temperate System
Viscosimètres Viscosimètres rotatifs
The ROTATEMP high-temperate system is designed to be used with the IKA ROTAVISC viscometers.
  • Fast and reliable heating: Thanks to the proven heating technology of IKA magnetic stirrers, your sample will heat up to 300 °C in only 16 minutes (using a silicone oil) without overshooting
  • Save valuable lab space: Heating oven and control unit are combined in one device with a small footprint
  • Automate, control, document: ROTATEMP can be connected to the viscometer software labworldsoft® 6 Visc
It reaches up to 300 °C and enables precise temperature control of highly viscous media that are difficult to measure, such as asphalt, bitumen, wax, polymers, candy base or hot melt adhesives.
Information de livraison: Package includes ROTAT extraction tool, ROTAT-C sample chamber, ROTAT-D-C disposable sample chamber (5 pcs.), alignment bracket, VOLS 1.11 and ROTAT rack.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité