Recirculating chiller, environmentally friendly, RC 2 lite
Fournisseur: IKA
25006624
IKAA25006624EA 3000 EUR InStock
IKAA25006624
Recirculating chiller, environmentally friendly, RC 2 lite
Thermostats à circulation Systèmes à circulation fermée Circulateurs de refroidissement
Compact recirculating chiller with powerful 400 W cooling capacity, designed for simple cooling tasks down to –10 °C. The RC 2 lite is the perfect peripheral device for the cooling of rotary evaporators, reflux condensers or as a cooling source for devices that dissipate heat with the help of a heat exchanger (double-walled grinding containers, cooled incubators, laboratory reactors, cooling coils). The easily accessible and easy-to-clean mesh filter along with low filling volume of just one litre enables fast cooling. Thanks to the natural refrigerant R290, the RC 2 lite is remarkably environmentally friendly.
  • High cooling capacity
  • Faster cooling
  • Enables cooling of a high number of external applications without refill of liquids
  • The filling level in the reservoir can be clearly observed through the large, illuminated sight glass
  • The powerful pressure and suction pump enables the simultaneous operation of several small applications as well as external applications in open bath vessels in combination with a level controller
  • Environmentally friendly refrigerant
The device is ideally suited for numerous applications in laboratories, such as cooling rotary evaporators, reflux condensers or as a cooling source for devices that dissipate heat with the help of a heat exchanger (double-walled grinding containers, cooled incubators, laboratory reactors, cooling coils).
Max. flow rate / max. pressure: 18 L/min, 0,15 (pressure) 0,35 bar (suction).
Certifications: IP protection class according to DIN EN 60529: IP 21.
Attention: Cooling capacity at +20; +10; 0; –10 °C.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité