Vacuum pumps, for RV 10 evaporators
Fournisseur: IKA
3980000 20016236
IKAA3980000EA 2340 EUR InStock
IKAA3980000 IKAA20016236
Vacuum pumps, for RV 10 evaporators
Pompe Pompes à vide pour évaporateurs rotatifs
LVS 105T 10-ef: Compact, dry running, chemical resistant pump systems, used for many applications in chemical laboratories and research.
MVP 10 basic:
Slimline and compact design dry diaphragm pump that is highly resistant to chemicals as all parts that come into contact with gas are made of PTFE or PTFE compounds and oil-free.
VACSTAR digital:
The four-chamber membrane vacuum pump convinces with its high suction capacity, low space requirement and high serviceability. The setting of the parameters is carried out through a rotary-push button over a digital display showing current rpm.More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité