Dry vacuum and chemical resistant diaphragm pumps (MPC)
Fournisseur: Welch by Gardner Denver
412522 412543 412422-02 412443-02 412521 412781 412782 412922 412783 412983 412422-10 412443-10 412421-02 412021 415722
ILMV412522EA 1290 EUR InStock
ILMV412522 181-0515 ILMV412422-02 ILMV412443-02 ILMV412521 ILMV412781 ILMV412782 ILMV412922 ILMV412783 ILMV412983 ILMV412422-10 ILMV412443-10 ILMV412421-02 ILMV412021 ILMV415722
Dry vacuum and chemical resistant diaphragm pumps (MPC)
Pompe Pompes à vide à membrane
The MPC chemical duty diaphragm pumps use PTFE and other fluorinated plastics for the wetted parts to allow aggressive solvent and acid vapours to be pumped. If the pump should handle harsh chemical vapours, then the superior chemical resistant X2 version with higher grade seals and thicker coatings are the best choice. These are well known for quiet operation and running smoothness.
  • Different formats are available (1, 2, 4 head diaphragm pumps)
  • Low weight and compact design
  • Can be moved to different locations based on applications
They are available in one or multistage configuration with controlled (ecoflex) or uncontrolled pumping speed. Ecoflex diaphragm pumps are equipped with a state of the art vacuum control which allows the pumping speed to be varied. The vacuum can be adjusted precisely to individual needs with the result that the process runs more efficiently and is reproducible. The additional pressure reduction allows the distillation of mixtures almost automatically and without fractionated operation. Ecoflex diaphragm pumps guarantee a positive economic and ecological balance with the result that operation costs can be reduced by up to 80%.
Applications include rotary evaporation, vacuum oven, industrial applications, desiccation, distillation, filtration, gel drying, concentrator, gas sampling, drying and degassing.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
SERVICES
Services: Besoin d'une installation, d'un IQOQ, d'un entretien ou d'une réparation? Contactez-nous : technical.service.be@vwr.com
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité