tempmate.®-M1 Multi-Use Temperature Data Logger
Fournisseur: tempmate
200100
IMEC200100EA 50 EUR InStock
IMEC200100
tempmate.®-M1 Multi-Use Temperature Data Logger
Enregistreurs de données
Intuitive multi-use temperature logger for reliable low-cost cold chain monitoring. For temperature recording with internal or external temperature sensor, multifunction display and automatic PDF report generation.
  • Product-related temperature monitoring along the entire cold chain, in warehouses or fridges
  • Cost-effective and reliable
  • Smart design, intuitive operation
  • Waterproof and food compatible
  • Free available software for individual logger configuration
  • Detailed PDF report including graph and table
  • Multifunction display with battery indicator
tempmate.®-M1 loggers are specially designed for temperature monitoring in pharmaceutical cold chain logistics. As preprogrammed device tempmate.®-M1 can be instantly used to control your temperature sensitive consignment. The PDF report created contains detailed information such as data curve, every single measurement as a table and statistical information.
With the optional free available tempbase.®2 software, you can ensure individual logger configuration and detailed data analysis or data management is also possible. Moreover, the device does not require labeling per IATA's Dangerous Goods Regulations.
Certifications: CE, EN 12830, RoHs, Validation certificate, MSDS 2023.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité