tempmate.®-i1 Single Use Indicator, 15 to 25 °C No Start Delay, No Alarm Delay
Fournisseur: tempmate
I1CS300
IMECI1CS300EA 220 EUR InStock
IMECI1CS300
tempmate.®-i1 Single Use Indicator, 15 to 25 °C No Start Delay, No Alarm Delay
Enregistreurs de données
Single use temperature indicator for reliable low-cost cold chain monitoring.
  • Product-related temperature monitoring along the entire cold chain
  • Cost-effective and reliable
  • Smart design, intuitive operation
  • Waterproof and food compatible
  • No additional hardware or software required
  • Storage time 4 years
  • Intelligent LEDs for status monitoring
tempmate.®-i1 indicators are specially designed for temperature monitoring in pharmaceutical cold chain logistics. As preprogrammed device tempmate.®-i1 can be instantly used to control your temperature sensitive consignment. By comparison to chemical indicators, the 3 LEDs at the frontside of the electronic indicator gives you a fast and clear feedback of your goods condition.
Certifications: MSDS, CE.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité