tempmate® S1 Single Use Temperature Data Logger, 15 min, L1 8 °C, H1 25 °C, SD 60 min
Fournisseur: tempmate
S1C15A79100
IMECS1C15A79100EA 0 EUR InStock
IMECS1C15A79100
tempmate® S1 Single Use Temperature Data Logger, 15 min, L1 8 °C, H1 25 °C, SD 60 min
Enregistreurs de données
Pre-programmed. Outstanding single-use temperature logger for reliable cold chain monitoring. Integrated USB port and PDF and CSV generating software on-board.
  • Pre-programmed
  • Cost-effective and reliable
  • Smart design, intuitive operation
  • Waterproof and food grade packing
  • No additional hardware or software required
  • Detailed PDF and CSV report including graph and table
  • Intelligent LEDs for status monitoring
  • Extended shelf-life of two years
tempmate.®-S1 loggers are specially designed for temperature monitoring in any cold chain logistics. As a pre-programmed device tempmate.®-S1 can be instantly used to monitor your temperature sensitive consignment. The automatically created PDF and CSV report, containing detailed information such as data curve, every single measurement as a table and statistical information. The optional tempbase.® software for data management and reprogramming can be downloaded for free. Moreover, the device does not require labeling per IATA's dangerous goods regulations.
Certifications: EN 12830, CE, RoHS.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité