tempmate.®-C1 Dry Ice Temperature Data Logger
Fournisseur: tempmate
TC1-000
IMECTC1-000EA 75 EUR InStock
IMECTC1-000
tempmate.®-C1 Dry Ice Temperature Data Logger
Enregistreurs de données
Single-use dry-ice monitoring solution for direct measurements in ultra-low temperatures without accessories.
  • Pre-programmed
  • 90 days active runtime (restartable)
  • Cost-effective and reliable
  • Smart design, intuitive operation
  • Waterproof and food grade packing
  • No additional hardware required
  • Detailed PDF and CSV report including graph and table
  • Big LED Display for status monitoring
  • Extended shelf-life of two years
Your tempmate.®-C1 can record temperatures as low as −90 °C and thus even provide a reliable overview of your dry ice transports. For this purpose, the entire device is developed in such a way that every single component, be it the electronics or the housing, can withstand extreme temperatures from −90 to +70 °C without any problems. The tempmate.®-C1 is fully configurable by the customer, allowing you to tailor it to your exact application. Start and stop modes, log interval and alarms, the device adapts to your needs and requirements. The 24 months shelf-life and up to 90 days runtime offer you a lot of freedom in scheduling your resources. Are you planning multiple deployments? Within the maximum of active 90 days, you can start and stop your tempmate.®-C1 multiple times.
Certifications: CE, RoHs, EN 12830, RTC-DO160.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité