Anti-IgG Chicken Polyclonal Antibody (Biotin (NHS-LC))
Fournisseur: ImmunoReagents
IR1021
IMMRIR1021EA 169 EUR InStock
IMMRIR1021
Anti-IgG Chicken Polyclonal Antibody (Biotin (NHS-LC))
Anticorps
ImmunoReagents’ Biotinylated antibody conjugates are widely used due to their ability to enhance antibody signal in most immunoassays. Biotinylated secondary antibodies are used in most immunoassay formats (i.e., ELISA, Immunohistochemistry (IHC), and Western blotting). While Biotinylated antibodies by themselves do not produce a signal that can be visualised, biotin has a high affinity and specificity to Streptavidin/Avidin. Using ImmunoReagents’ streptavidin conjugates with biotinylated antibodies results in signal amplification.
  • Origin: US
ImmunoReagents is a manufacturer ofMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité