Anti-IgG Donkey Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fournisseur: ImmunoReagents
IR1181
IMMRIR1181EA 186 EUR InStock
IMMRIR1181
Anti-IgG Donkey Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorps
ImmunoReagents’ FITC Conjugates are used in immunofluorescence microscopy, flow cytometry, FLISA (fluorescent ELISA) and high throughput screening assays. Fluorescein is typically excited by the 488 nm line of an argon laser, and emission is collected at 518 nm. FITC antibody conjugates are yellow-orange in color but emit a yellow-green color upon Em 518 nm. FITC antibody conjugates are sensitive to light. The typical FITC to antibody ratio (F/P) is >4.0 Excitation/Emission = 494 nm / 518 nm Emission Color = Green (Similar Dyes: Alexa Fluor 488, Cy2, DyLight ® 488)
  • Origin: US
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité