Anti-IgG Donkey Polyclonal Antibody (DyLight® 633)
Fournisseur: ImmunoReagents
IR1317
IMMRIR1317EA 135 EUR InStock
IMMRIR1317
Anti-IgG Donkey Polyclonal Antibody (DyLight® 633)
Anticorps
The ImmunoReagents' DyLight® 633 conjugates fluoresces red with physical properties comparable to other 633 dyes. DyLight® 633 is ideal for multi-Colour applications. DyLight® 633 is available as reactive labeling agents and as conjugates of secondary antibodies and biotin-binding proteins for use in fluorescence microscopy, flow cytometry, Western blotting, ELISA, high-content screening and other array platforms. Excitation/Emission = 638 nm/658 nm Emission Colour = Red (Similar Dyes: Alexa Fluor 633).
  • Origin: US
ImmunoReagents' antibodies are suitable for most immunoassayMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité