Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Fournisseur: ImmunoReagents
IR2126
IMMRIR2126EA 222 EUR InStock
IMMRIR2126
Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticorps
ImmunoReagents’ Alkaline Phosphatase (AP) antibody conjugates are used in ELISA, Immunohistochemistry (IHC) and Western blotting techniques. AP is a 140 kDa enzyme that changes colour in the presence of PNPP or BCIP/NBT substrates. These conjugates have a linear reaction rate thus allowing the researcher to increase sensitivity by prolonging substrate incubation times.
  • Origin: US
ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.
Products will beMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité