Anti-IgG Goat Polyclonal Antibody (DyLight® 800)
Fournisseur: ImmunoReagents
IR2173
IMMRIR2173EA 251 EUR InStock
IMMRIR2173
Anti-IgG Goat Polyclonal Antibody (DyLight® 800)
Anticorps
The ImmunoReagents' DyLight® 800 conjugates uses a near-IR fluorphore that is invisible to the naked eye but increases the staining options when using infrared imaging systems. DyLight® 800 has spectral properties that are very similar to other near-IR dyes, including Alexa Fluor 790 and IRDye 800, and has high solubility for maximal incorporation of the dye into the biomolecule of interest. Excitation/Emission = 777 nm/794 nm Emission Colour = Near IR (Similar Dyes: Alexa Fluor 790, IRDye 800)
  • Origin: US
ImmunoReagents' antibodies are suitable for most immunoassay formats andMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité