dataLink pro Petri Dish Labelling Solution
Fournisseur: INTERSCIENCE
439050
INSC439050EA 17220 EUR InStock
INSC439050
dataLink pro Petri Dish Labelling Solution
Imprimantes Label Printers
dataLink pro is designed for the identification of Petri dishes during its microbiological analysis. The dataLink software controls and transmits data on a label.
  • User account management
  • Monitoring of sample data and automation of settings
  • Label size: 50×5 mm
  • Audit trail, 21 CFR part 11
  • Semi-automatic labelling platform
Semi-automated labelling on the edge of the dish is great for large production analyses. With a wave of the hand, the label is applied to the edge of the Petri dish.
Controlled via the dataLink software, all information relating to the analysis data is traced and sent to the printer. There are three label formats alphanumeric, barcode and datamatrix.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité