Micro-90 Cleaning Solution
Fournisseur: ISMATEC PART OF COLE PARMER
18100-05
ISMC18100-05EA 47.3 EUR InStock
ISMC18100-05
Micro-90 Cleaning Solution
Produits nettoyants Détergents et Nettoyants de Surfaces
This concentrated cleaning solution for critical applications. Solution cleans glass, most metals, plastic, rubber, fabric, and is safe for all your ultrasonic cleaners.
  • Radioactive contaminants will not reduce cleaning capability
  • Solution works at any concentration and rinses clean
  • Most applications require only 1,25 to 2,5 fluid ounces per gallon of water
Micro-90 is a mild, yet powerful, free-rinsing cleaner designed for use in critical cleaning and general-purpose applications. This environmentally friendly solution contains no phosphates, CFCs, ODCs, or borates. It safely replaces chromic acid to remove protein from glassware. Designed for use in CIP, ultrasonic, and immersion applications. Can be used in validated processses. A 1 to 2% solution is effective in most applications. Typical pH: 9,5. NSF-Registered for USDA A1 use.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité