Synonyms:
1-((2-Chlorophenyl)diphenylmethyl)-1H-pyrazole, TRAM-34
J60019.MB
J60019.MBEA
306
EUR
InStock
J60019.MB
TRAM 34
TRAM 34
CAS No.: 289905-88-0
Molecular Formula: C22H17ClN2
Formula Weight: 344.84
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Yellow
Melting Point: 145-147°
Solubility: Soluble in DMSO at 2mg/ml. Insoluble in water
MDL No.: MFCD09842562
Applications: TRAM-34 is a potent inhibitor of the intermediate-conductance calcium-activated potassium channel