J60373.MF
J60373.MFEA
180
EUR
InStock
J60373.MF
Kaempferol ≥98%
Kaempferol
Purity: 98+%
CAS No.: 520-18-3
Molecular Formula: C15H10O6
Formula Weight: 286.24
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Melting Point: 276-278°
Solubility: Soluble in DMSO or ethanol to 100mM
Beilstein Reference: 304401
Merck Reference: 14,5274
MDL No.: MFCD00016938
Inhibits topoisomerase I catalyzed DNA religation. Induces apoptosis and is cytotoxic to human leukemic cell lines