J60574.22
J60574.22EA
31.9
EUR
InStock
J60574.22
2-Aminoazotoluene
2-Aminoazotoluene
CAS No.: 97-56-3
Molecular Formula: C14H15N3
Formula Weight: 225.29
Physical Form: Powder
Appearance: Orange brown
Odour: Odourless
Beilstein Reference: 6506005
Merck Reference: 14,420
MDL No.: MFCD00007733
C.I. 11160