J60678.MC
J60678.MCEA
122
EUR
InStock
J60678.MC
Lidocaine-N-ethyl bromide ≥99%
Lidocaine-N-ethyl bromide
Purity: 99+%
CAS No.: 21306-56-9
Molecular Formula: C16H27BrN2O
Formula Weight: 343.31
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline solid
Appearance: White
MDL No.: MFCD00083182
Applications: Blocker of voltage-activated sodium channels