Synonyms:
(S)-1-(2-Pyridyl)ethanol, (S)-1-(Pyridin-2-yl)ethanol, (S)-alpha-Methyl-2-pyridinemethanol
J60980.03
J60980.03EA
158
EUR
InStock
J60980.03
2-[(S)-1-Hydroxyethyl]pyridine
2-[(S)-1-Hydroxyethyl]pyridine
CAS No.: 59042-90-9
Molecular Formula: C7H9NO
Formula Weight: 123.16
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: Colourless
MDL No.: MFCD06795465
Applications: For synthesis of optically active products