J61132.03
J61132.03EA
48.1
EUR
InStock
J61132.03
Tropicamide ≥99%
Tropicamide
Purity: 99+%
CAS No.: 1508-75-4
Molecular Formula: C17H20N2O2
Formula Weight: 284.35
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Melting Point: 98°
MDL No.: MFCD00058580
Applications: A muscarinic antagonist