Synonyms:
alpha-(Aminomethyl)-4-hydroxybenzyl alcohol hydrochloride, 4-(2-Amino-1-hydroxyethyl)phenol hydrochloride, dl-Octopamine hydrochloride, 4-(2-amino-1-hydroxyethyl)phenol hydrochloride, Octopamine hydrochloride
J61281.09
J61281.09EA
125
EUR
InStock
J61281.09
(+/-)-Octopamine hydrochloride 99%
(+/-)-Octopamine hydrochloride
Purity: 99%
CAS No.: 770-05-8
Molecular Formula: C8H11NO2.xHCl
Formula Weight: 189.64
Physical Form: Powder
Appearance: White to off-white
Melting Point: 170° dec.
Beilstein Reference: 3915414
MDL No.: MFCD00012881
Applications: alpha-adrenoceptor agonist; invertebrate neurotransmitter and neuromodulator