Synonyms:
(-)-cis-3,4-Dihydro-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-1(2H)-benzopyran-3-yl Gallate, (2R,3R)-5,7-Dihydroxy-2-(3,4,5-trihydroxyphenyl)chroman-3-yl 3,4,5-trihydroxybenzoate, EGCG
J61745.MF
J61745.MFEA
179
EUR
InStock
J61745.MF
(-)-Epigallocatechin gallate
(-)-Epigallocatechin gallate
CAS No.: 989-51-5
Molecular Formula: C22H18O11
Formula Weight: 458.40
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Pale pink to pale orange
Melting Point: 216-218°
MDL No.: MFCD00075940
Applications: An antioxidant that acts as an anti-promoter of carcinogenesis, inhibits angiogenesis and induces apoptosis