Synonyms:
Halochondrine A potassium salt, OA potassium salt, 9,10-deepithio-9,10-didehydroacanthifolicin potassium salt
J61791.XS
J61791.XG
J61791.IX
J61791.XSEA
351
EUR
InStock
J61791.XS
J61791.XG
J61791.IX
Okadaic acid potassium salt 98%
Okadaic acid potassium salt
Potent inhibitor of serine/threonine-specific protein phosphatases 1 and 2A (PP1 and PP2A). Does not inhibit tyrosine or alkaline phosphatases