J61898.MA
J61898.MAEA
204
EUR
InStock
J61898.MA
Oligomycin
Oligomycin
CAS No.: 1404-19-9
Molecular Formula: C45H74O11
Formula Weight: 791.07
Physical Form: Powder
Appearance: White
Solubility: Sparingly soluble in water. Soluble in DMSO (20mg/ml) and ethanol (30mg/ml)
Beilstein Reference: 7615308
MDL No.: MFCD01779388
Applications: Inhibits respiration in mitochondria. Also inhibits ATP synthase