Synonyms:
1-(4-(tert-Butyl)phenyl)-4-(4-(hydroxydiphenylmethyl)piperidin-1-yl)butan-1-ol
J61936.MC
J61936.MCEA
267
EUR
InStock
J61936.MC
Terfenadine
Terfenadine
CAS No.: 50679-08-8
Molecular Formula: C32H41NO2
Formula Weight: 471.67
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to off-white
Melting Point: 145-152°
MDL No.: MFCD00079622
Applications: An antihistamine