J62097.03
J62097.03EA
228
EUR
InStock
J62097.03
1-(1-Naphthyl)piperazine hydrochloride
1-(1-Naphthyl)piperazine hydrochloride
CAS No.: 104113-71-5
Molecular Formula: C14H16N2.HCl
Formula Weight: 248.75
Physical Form: Solid
Appearance: Purple
MDL No.: MFCD00055161
Applications: 5-HT1 serotonin receptor agonist. 5-HT2 serotonin receptor antagonist