Synonyms:
TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
J62143.AP
J62143.APEA
241
EUR
InStock
J62143.AP
3,3',5,5'-Tétraméthylbenzidine en solution aqueuse, haute sensibilité prêt-à-l'emploi, précipitant
3,3',5,5'-Tétraméthylbenzidine
Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result.