Synonyms:
(R)-2-Oxothiazolidine-4-carboxylic acid, Procysteine
J62254.03
J62254.03EA
55.3
EUR
InStock
J62254.03
(R)-(-)-2-Oxothiazolidine-4-carboxylic acid
(R)-(-)-2-Oxothiazolidine-4-carboxylic acid
CAS No.: 19771-63-2
Molecular Formula: C4H5NO3S
Formula Weight: 147.15
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to off-white
Melting Point: 174° dec.
Solubility: Soluble in water
Beilstein Reference: 4179169
Merck Reference: 14,6950
MDL No.: MFCD00066092
Reagent for hepatic glutathione studies.