Synonyms:
Tetramethylrhodamine-5(6)-isothiocyanate, 5(6)-TRITC, MRITC, TRITC
J62258.MA
J62258.MAEA
239
EUR
InStock
J62258.MA
5(6)-Tetramethylrhodamine isothiocyanate
5(6)-Tetramethylrhodamine isothiocyanate [5(6)-TRITC]
CAS No.: 95197-95-8
Molecular Formula: C25H21N3O3S
Formula Weight: 443.52
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Light, Air and Moisture Sensitive
Appearance: Red to dark purple
Solubility: Soluble in DMSO or methanol
MDL No.: MFCD00063517
Applications: Reagent for fluorescent labelling of proteins