J62381.03
J62381.03EA
438
EUR
InStock
J62381.03
Etidronate disodium
Etidronate disodium
CAS No.: 7414-83-7
Molecular Formula: C2H6Na2O7P2
Formula Weight: 249.99
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline solid
Appearance: White
Melting Point: 300°
Merck Reference: 14,3863
MDL No.: MFCD00152567
A bisphosphonate bone resorption inhibitor