Synonyms:
(R)-1-(Pyridin-2-yl)ethanol
J62548.03
J62548.03EA
173
EUR
InStock
J62548.03
(R)-2-(1-Hydroxyethyl)pyridine
(R)-2-(1-Hydroxyethyl)pyridine
CAS No.: 27911-63-3
Molecular Formula: C7H9NO
Formula Weight: 123.16
Physical Form: Solid
Appearance: Colourless
Solubility: Soluble in ethanol
Beilstein Reference: 3537186
MDL No.: MFCD04972322
Applications: For synthesis of optically active products